missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Phenylhydrazono)malononitrile, TCI America™

Manufacturer:  TCI America P234625G

 View more versions of this product

Catalog No. P234625G



Specifications

Specifications

(Phenylhydrazono)malononitrile
133°C
306-18-3
MFCD00195983
MENUYOGJCXAFFU-UHFFFAOYSA-N
2-(phenylhydrazinylidene)propanedinitrile
67541
Crystal-Powder at 20°C
Yellow
25g
C9H6N4
3439
C1=CC=C(C=C1)NN=C(C#N)C#N
170.175
170.18
≥96.0% (GC)

Chemical Identifiers

306-18-3
170.175
MENUYOGJCXAFFU-UHFFFAOYSA-N
2-(phenylhydrazinylidene)propanedinitrile
C9H6N4
MFCD00195983
67541
C1=CC=C(C=C1)NN=C(C#N)C#N
Safety and Handling

Safety and Handling

TSCA : No

Documents