missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylhydrazinium Chloride, Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1072530250

 View more versions of this product

Catalog No. M1072530250



Description

Description

For analysis
Specifications

Specifications

Phenylhydrazinium Chloride
250g
C6H9ClN2
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
C1=CC=C(C=C1)NN.Cl
144.602
144.6
Reagent Ph. Eur.
Glass bottle
59-88-1
UN 2811
JOVOSQBPPZZESK-UHFFFAOYSA-N
phenylhydrazine;hydrochloride
60962
Solid

Chemical Identifiers

59-88-1
144.602
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
phenylhydrazine;hydrochloride
C6H9ClN2
JOVOSQBPPZZESK-UHFFFAOYSA-N
60962
C1=CC=C(C=C1)NN.Cl
Safety and Handling

Safety and Handling

H350: May cause cancer.
H341: Suspected of causing genetic defects.
H331: Toxic if inhaled.
H311: Toxic in contact with skin.
H301: Toxic if swallowed.
H372: Causes damage to organs through prolonged or repeated exposure.
H319: Causes serious eye irritation.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H400: Very toxic to aquatic life.

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 200-444-7

Documents
Promotions

Promotions