missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylhydrazine, TCI America™

Manufacturer:  TCI America P0183100G

 View more versions of this product

Catalog No. P0183100G



Specifications

Specifications

Phenylhydrazine
100g
C6H8N2
2572
HKOOXMFOFWEVGF-UHFFFAOYSA-N
phenylhydrazine
7516
108.14
≥98.0% (GC,T)
Yellow
100-63-0
MFCD00007573
hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
C1=CC=C(C=C1)NN
108.144
CHEBI:27924
Clear Liquid at 20°C

Chemical Identifiers

100-63-0
108.144
HKOOXMFOFWEVGF-UHFFFAOYSA-N
7516
phenylhydrazine
C6H8N2
MFCD00007573
hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
CHEBI:27924
C1=CC=C(C=C1)NN
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0470

RTECSNumber : MV8925000

TSCA : Yes

Documents