Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylhydrazine hydrochloride, 99%

Chemical Identifiers

Chemical Name or Material Phenylhydrazine hydrochloride
CAS 59-88-1
Molecular Formula C6H8N2·HCl
Formula Weight 144.61
MDL Number MFCD00012924
UN Number UN2811
Beilstein 3594958
Merck Index 14,7293
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1464522

  SDS
alfa aesar™
A1464522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.61
CAS 59-88-1
Chemical Name or Material Phenylhydrazine hydrochloride
TSCA TSCA
Assay 99%
Molecular Formula C6H8N2·HCl
MDL Number MFCD00012924
UN Number UN2811
Beilstein 3594958
Merck Index 14,7293
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H350-H372-H341-H315-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Melting Point ≈252°C (dec.)
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number MV9000000
Quantity 100g

Applications
Phenylhydrazine hydrochloride is used as a selective mannosidase inhibitor. It is used in the study of tyrosine phosphorylation of janus protein tyrosine kinase in the EPO-responsive normal erythroblastoid cells of anemic mice. It is used in steroid assays and as an N-protecting reagent. Its free base, phenylhydrazine is used to prepare indoles, which find application as intermediates in the synthesis of various dyes and pharmaceuticals.

Solubility
Soluble in water and alcohol.

Notes
Air and light sensitive. Hygroscopic. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H350-H372-H341-H315-H319-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.