Learn More
Phenylglyoxal monohydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1067722
Description
Phenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylglyoxal monohydrate was used to modify pig muscle carbonic anhydraseIII and as a specific reagent for arginine groups. It was also used to prepare pyrrolinone and furan derivatives and as as a chemiluminescent reagent for the determination of purines.
Solubility
Partly miscible with water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Phenylglyoxal monohydrate | |
71°C to 77°C | |
C8H8O3 | |
100 g | |
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone | |
NBIBDIKAOBCFJN-UHFFFAOYSA-N | |
2,2-dihydroxy-1-phenylethanone | |
99611 | |
97% |
1075-06-5 | |
142°C (125 mmHg) | |
MFCD00149499 | |
1854721 | |
Partly miscible with water. | |
C1=CC=C(C=C1)C(=O)C(O)O | |
152.149 | |
152.15 (134.12 Anhydrous) |
Chemical Identifiers
1075-06-5 | |
152.149 | |
NBIBDIKAOBCFJN-UHFFFAOYSA-N | |
99611 | |
C1=CC=C(C=C1)C(=O)C(O)O |
C8H8O3 | |
MFCD00149499 | |
dihydroxyacetophenone, acetophenone, dihydroxy, dihydroxy 1-phenylethanone, 2,2-dihydroxy-1-phenyl-ethanone, ethanone, 2,2-dihydroxy-1-phenyl, acetophenone, 2,2-dihydroxy, alpha,alpha-dihydroxyacetophenone, dioxyacetophenone, alpha,alpha-dihydroxy-acetophenone | |
2,2-dihydroxy-1-phenylethanone |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 214-036-1
RTECSNumber : KM5775180
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only