Please login to your online account to display your discounted pricing

α-Phenylcinnamic Acid 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  130421000

 View more versions of this product

Catalog No. AC130421000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 224.26
Physical Form Crystalline Powder
Quantity 100g
Color Almost white
TSCA TSCA
EINECS Number 202-069-4
Melting Point 171° to 175°C
Merck Index 15, 7393
Beilstein 09, 691
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula C6H5CH=C(C6H5)CO2H
CAS 91-48-5
Chemical Name or Material α-Phenylcinnamic acid
MDL Number MFCD00004252
Name Note 98%
Assay Percent Range 98%
Molecular Formula C15H12O2

Specifications

Chemical Name or Material α-Phenylcinnamic acid
Name Note 98%
CAS 91-48-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H12O2
Linear Formula C6H5CH=C(C6H5)CO2H
Formula Weight 224.26
MDL Number MFCD00004252