missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Phenylcinnamic Acid 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 130420250

 View more versions of this product

Catalog No. AC130420250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
224.26
Almost white
15, 7393
100.0
Authentic
α-Phenylcinnamic acid
91-48-5
98%
C6H5CH=C(C6H5)CO2H
Crystalline Powder
25g
09, 691
171° to 175°C
97.5
Glass bottle
98%
C15H12O2
MFCD00004252

Chemical Identifiers

91-48-5
100.0
C15H12O2
224.26
α-Phenylcinnamic acid, 98%
97.5
98%
C6H5CH=C(C6H5)CO2H
MFCD00004252
≥97.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 202-069-4

TSCA : TSCA