missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Phenylcinnamic Acid 98%, ACROS Organics™

Chemical Identifiers

CAS 91-48-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H12O2
Linear Formula C6H5CH=C(C6H5)CO2H
Formula Weight 224.26
MDL Number MFCD00004252
Chemical Name or Material α-Phenylcinnamic acid, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC130421000 SDS Thermo Fisher Scientific
130421000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC130420250 SDS Thermo Fisher Scientific
130420250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

91-48-5
100.0
C15H12O2
224.26
α-Phenylcinnamic acid, 98%
97.5
98%
C6H5CH=C(C6H5)CO2H
MFCD00004252
≥97.5%

Specifications

224.26
100g
171° to 175°C
09, 691
97.5
Authentic
91-48-5
98%
α-Phenylcinnamic acid, 98%
Crystalline Powder
Almost white
15, 7393
100.0
Glass bottle
C6H5CH=C(C6H5)CO2H
MFCD00004252
C15H12O2
≥97.5%

Safety and Handling

EINECSNumber : 202-069-4

TSCA : TSCA