missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylbutazone, 99+%, ACROS Organics™

Chemical Identifiers

CAS 50-33-9
Molecular Formula C19H20N2O2
Molecular Weight (g/mol) 308.38
MDL Number MFCD00005500
InChI Key VYMDGNCVAMGZFE-UHFFFAOYSA-N
Synonym butacote, butadion, butapirazol, butapyrazole, butazolidin, butazolidine, diphenylbutazone, fenilbutazona, phenylbutazone, phenylbutazonum
PubChem CID 4781
ChEBI CHEBI:48574
IUPAC Name 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
SMILES CCCCC1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199310050
SDS View Documents
Thermo Fisher Scientific
199310050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

50-33-9
308.38
VYMDGNCVAMGZFE-UHFFFAOYSA-N
4781
4-butyl-1,2-diphenylpyrazolidine-3,5-dione
C19H20N2O2
MFCD00005500
butacote, butadion, butapirazol, butapyrazole, butazolidin, butazolidine, diphenylbutazone, fenilbutazona, phenylbutazone, phenylbutazonum
CHEBI:48574
CCCCC1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Authentic
White
5g
99+%
MFCD00005500
butacote, butadion, butapirazol, butapyrazole, butazolidin, butazolidine, diphenylbutazone, fenilbutazona, phenylbutazone, phenylbutazonum
VYMDGNCVAMGZFE-UHFFFAOYSA-N
4-butyl-1,2-diphenylpyrazolidine-3,5-dione
4781
308.38
99+%
Glass bottle
104°C to 107°C
50-33-9
C19H20N2O2
15, 7390
Solubility in water: slightly soluble. Other solubilities: soluble in solutions of alkali hydroxides,freely soluble in acetone,ether and ethyl acetate
CCCCC1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3
308.38
CHEBI:48574
Powder
Phenylbutazone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-029-