Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylbutazone, 98%

Chemical Identifiers

Chemical Name or Material Phenylbutazone
CAS 50-33-9
Molecular Formula C19H20N2O2
Formula Weight 308.38
MDL Number MFCD00005500
UN Number UN2811
Beilstein 290080
Merck Index 14,7277
Synonym 4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0344922

  SDS
alfa aesar™
L0344922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 308.38
Chemical Name or Material Phenylbutazone
CAS 50-33-9
Synonym 4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione
Health Hazard 2 GHS H Statement
H301-H351-H361-H319
Toxic if swallowed.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
Molecular Formula C19H20N2O2
Merck Index 14,7277
MDL Number MFCD00005500
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Quantity 100g
RTECS Number UQ8225000
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Beilstein 290080
Melting Point 105° to 108°C
Health Hazard 1 Danger
TSCA TSCA
Odor Odorless
Assay 98%