missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130360500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenylboronic acid | |
96.5 | |
217°C to 222°C | |
Authentic | |
Glass bottle | |
C6H5B(OH)2 | |
50 g | |
01,833; 02,317; 03,221; 05,513; 07,284; 09,23; 11,408; 15,254 | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
phenylboronic acid | |
66827 | |
121.93 | |
Crystalline Powder |
98-80-6 | |
100.0 | |
White | |
98% min. (HPLC) | |
C6H7BO2 | |
MFCD00002103 | |
16, 920 | |
15, 1070 | |
Solubility in water: 10g/L (20°C). Other solubilities: solubility in benzene : 1.75%, xylene : 1.2%, methanol : 178%, ether : 30.2% | |
OB(O)C1=CC=CC=C1 | |
121.93 | |
CHEBI:44923 | |
98+% |
Chemical Identifiers
98-80-6 | |
121.93 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
66827 | |
phenylboronic acid |
C6H7BO2 | |
MFCD00002103 | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
CHEBI:44923 | |
OB(O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 202-701-9
RUO – Research Use Only