Please login to your online account to display your discounted pricing

Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis, ACROS Organics™

$69.40 - $69.40

Chemical Identifiers

Chemical Name or Material Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis
CAS 538-62-5
Molecular Formula C13H12N4O
Linear Formula C6H5N=NCONHNHC6H5
Formula Weight 240.26
MDL Number MFCD00003024
Synonym Diphenylcarbazone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154450100

  SDS
thermo fisher scientific
AC154450100
Glass bottle 10g Each for $69.40

AC154450500

  SDS
thermo fisher scientific
AC154450500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 240.26
Quantity 10g
Color Orange
EINECS Number 208-698-0
TSCA TSCA
RTECS Number LQ9420000
Melting Point 153° to 157°C
Infrared Spectrum Authentic
Chemical Name or Material Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis
Packaging Glass bottle
Sulfated Ash 0.1% max.
Suitability suitable for determination of mercury
Molecular Formula C13H12N4O
Linear Formula C6H5N=NCONHNHC6H5
MDL Number MFCD00003024
CAS 538-62-5
Synonym Diphenylcarbazone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable solid.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Indicator

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable solid.