Please login to your online account to display your discounted pricing

Phenylarsonic acid, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224061000

 View more versions of this product

Catalog No. AC224061000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 202.04
Quantity 100g
Color White to slightly yellow
Merck Index 15, 1069
EINECS Number 202-631-9
TSCA TSCA
RTECS Number CY3150000
Packaging Glass bottle
Synonym Benzenearsonic acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Infrared Spectrum Authentic
Chemical Name or Material Phenylarsonic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H7AsO3
Linear Formula C6H5AsO3H2
MDL Number MFCD00002097
CAS 98-05-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Phenylarsonic acid
Name Note 97%
CAS 98-05-5
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H7AsO3
Linear Formula C6H5AsO3H2
Formula Weight 202.04
MDL Number MFCD00002097
Synonym Benzenearsonic acid
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents