missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylarsonic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 224060250

 View more versions of this product

Catalog No. AC224060250


 

Specifications Description & Specifications

Specifications

Phenylarsonic acid
25g
98-05-5
Glass bottle
≥96.0%
C6H7AsO3
202.04
15, 1069
White to slightly yellow
97%
Authentic
Crystalline Powder
97%
C6H5AsO3H2
MFCD00002097
Benzenearsonic acid

Chemical Identifiers

98-05-5
C6H7AsO3
202.04
Benzenearsonic acid
≥96.0%
97%
C6H5AsO3H2
MFCD00002097
Phenylarsonic acid, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-631-9

RTECSNumber : CY3150000

TSCA : TSCA