Please login to your online account to display your discounted pricing

Phenylarsonic acid, 97%, ACROS Organics™

$196.11 - $519.43

Chemical Identifiers

Chemical Name or Material Phenylarsonic acid
Name Note 97%
CAS 98-05-5
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H7AsO3
Linear Formula C6H5AsO3H2
Formula Weight 202.04
MDL Number MFCD00002097
Synonym Benzenearsonic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC224060250

  SDS
thermo fisher scientific
224060250
Glass bottle 25g Each for $196.11

AC224061000

  SDS
thermo fisher scientific
224061000
Glass bottle 100g Each for $519.43
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 202.04
Quantity 25g
Color White to slightly yellow
EINECS Number 202-631-9
TSCA TSCA
RTECS Number CY3150000
Merck Index 15, 1069
Chemical Name or Material Phenylarsonic acid
Packaging Glass bottle
Name Note 97%
Molecular Formula C6H7AsO3
Linear Formula C6H5AsO3H2
CAS 98-05-5
Synonym Benzenearsonic acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00002097
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.