Please login to your online account to display your discounted pricing

Phenylarsine Oxide, Reagent Grade, LabChem™

$246.03 - $246.03

Chemical Identifiers

Chemical Name or Material Phenylarsine Oxide
CAS 637-03-6
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C6H5AsO
Formula Weight 168.03
UN Number UN1557
Synonym PAO
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC183357

 
lab chem
LC183357
Amber Glass 10g Each for $246.03
Description & Specifications

Specifications

Formula Weight 168.03
Solubility Information Soluble in water
Decomposition Information Carbon dioxide; Carbon monoxide; Arsenic and its oxides
Health Hazard 1 Danger
EINECS Number 211-275-3
Chemical Name or Material Phenylarsine Oxide
CAS Max % 100
Purity Grade Reagent Grade
Quantity 10g
Health Hazard 2 GHS H Statement
Toxic if swallowed or if inhaled.
Very toxic to aquatic life with long lasting effects.
Linear Formula C6H5AsO
Identification Passes Test
Storage Information Room Temperature
Melting Point 145°C
CAS 637-03-6
Synonym PAO
Physical Form Powder
Packaging Amber Glass
UN Number UN1557
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Color White
Safety and Handling

Danger

GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed or if inhaled.
Very toxic to aquatic life with long lasting effects.