Please login to your online account to display your discounted pricing

Phenylarsine Oxide, For BOD, Certified, 0.0375N ±0.0005N (0.01875M), LabChem™

$239.20 - $239.20

Chemical Identifiers

Chemical Name or Material Phenylarsine Oxide
CAS 7732-18-5
CAS Max % 98.92
CAS 7647-01-0
CAS Max % 0.4
CAS 1310-73-2
CAS Max % 0.35
CAS 637-03-6
CAS Max % 0.33
Name Note For BOD
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC183902

 
lab chem
LC183902
1L Each for $239.20
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 168.03
Chemical Name or Material Phenylarsine Oxide
Traceability to NIST Traceable to NIST
CAS Max % 0.35
Decomposition Information Arsenic and its oxides; Carbon monoxide; Carbon dioxide
CAS 637-03-6
Color Clear, colorless to slight yellow
pH 7.0 to 9.0
Quantity 1L
CAS Max % 0.33
Name Note For BOD
Physical Form Liquid
Concentration 0.0375N ±0.0005N (0.01875M)
Linear Formula C6H5AsO
CAS 7732-18-5
Synonym PAO
Health Hazard 1 Warning
CAS Max % 98.92
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Identification Passes Test
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Amber Glass
CAS Max % 0.4
Solubility Soluble in water
CAS 1310-73-2
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.