missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylarsine Oxide, Certified, 0.025N ±0.001N (0.0125M), LabChem™

Manufacturer:  LabChem LC183802

 View more versions of this product

Catalog No. LC183802



Specifications

Specifications

Phenylarsine Oxide
Amber Glass
1L
637-03-6,7732-18-5
C6H5AsO
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
Soluble in water
C1=CC=C(C=C1)[As]=O
168.027
CHEBI:75253
Liquid
Passes Test
Colorless
6.0 to 7.0
99.9,0.1
C6H5AsO
0.025N ±0.001N (0.0125M)
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778
168.03
Certified

Chemical Identifiers

637-03-6
168.027
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
C1=CC=C(C=C1)[As]=O
C6H5AsO
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778
arsorosobenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

Documents