missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylarsine oxide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 180350010

 View more versions of this product

Catalog No. AC180350010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenylarsine oxide
145°C to 148°C
Authentic
Glass bottle
C6H5As=O
1 g
15, 7048
Solubility in water: insoluble. Other solubilities: very soluble in benzene,slightly soluble in ethanol,insoluble in ether
O=[As]C1=CC=CC=C1
168.03
CHEBI:75253
97%
637-03-6
White
97%
C6H5AsO
MFCD00001990
16, I, 438
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778
168.03
Crystalline Powder or Crystals

Chemical Identifiers

637-03-6
168.03
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778
arsorosobenzene
C6H5AsO
MFCD00001990
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
O=[As]C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

GHS Signal Word: Danger

EINECSNumber : 211-275-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only