missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylarsine oxide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1467414

 View more versions of this product

Catalog No. AAA1467414



Description

Description

Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.

Solubility
Soluble in dimethyl sulfoxide, chloroform, benzene, ether, ethanol, 50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0 mg/ml) and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Phenylarsine oxide
140°C to 160°C
MFCD00001990
UN3465
14,6949
Soluble in dimethyl sulfoxide,chloroform,benzene,ether,ethanol,50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0mg/ml) and toluene. Insoluble in water.
O=[As]C1=CC=CC=C1
168.03
CHEBI:75253
97%
637-03-6
C6H5AsO
25 g
2935227
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778
168.03

Chemical Identifiers

637-03-6
168.03
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778
arsorosobenzene
C6H5AsO
MFCD00001990
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
O=[As]C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P261-P264-P270-P271-P301+P310-P304+P340-P311-P330-P501c

H301+H331

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 211-275-3

RTECSNumber : CH8100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only