Learn More
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1467406
Description
Phenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylarsine oxide is used as a poultry coccidiostat in veterinary medicine. It is also used in a membrane-permeable protein tyrosine phosphatase inhibitor.
Solubility
Soluble in dimethyl sulfoxide, chloroform, benzene, ether, ethanol, 50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0 mg/ml) and toluene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Phenylarsine oxide | |
140°C to 160°C | |
MFCD00001990 | |
UN3465 | |
14,6949 | |
Soluble in dimethyl sulfoxide,chloroform,benzene,ether,ethanol,50:50 dimethyl sulfoxide:1 M hydrochloric acid (3.0mg/ml) and toluene. Insoluble in water. | |
O=[As]C1=CC=CC=C1 | |
168.03 | |
CHEBI:75253 | |
97% |
637-03-6 | |
C6H5AsO | |
5 g | |
2935227 | |
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
arsorosobenzene | |
4778 | |
168.03 |
Chemical Identifiers
637-03-6 | |
168.03 | |
BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
4778 | |
arsorosobenzene |
C6H5AsO | |
MFCD00001990 | |
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
CHEBI:75253 | |
O=[As]C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P264-P270-P271-P301+P310-P304+P340-P311-P330-P501c
H301+H331
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 211-275-3
RTECSNumber : CH8100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only