Please login to your online account to display your discounted pricing

Phenylacetaldehyde Dimethyl Acetal 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130275000

 View more versions of this product

Catalog No. AC130275000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 166.22
Quantity 500g
Color Clear colorless to pale yellow
Beilstein 07, 293
EINECS Number 202-945-6
TSCA TSCA
Specific Gravity 1.002 to 1.006 (25°/25°C)
RTECS Number AB3040000
Density 1.004g/mL
Acid Value 1mg KOH/g max.
Chemical Name or Material Phenylacetaldehyde dimethyl acetal
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H5CH2CH(OCH3)2
MDL Number MFCD00008487
CAS 101-48-4
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Phenylacetaldehyde dimethyl acetal
Name Note 98%
CAS 101-48-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H14O2
Linear Formula C6H5CH2CH(OCH3)2
Formula Weight 166.22
MDL Number MFCD00008487