missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 130271000
Specifications
Phenylacetaldehyde dimethyl acetal | |
101-48-4 | |
Colorless to Yellow | |
219°C to 221°C (754.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4930 to 1.495 | |
MFCD00008487 | |
07, 293 | |
2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal | |
WNJSKZBEWNVKGU-UHFFFAOYSA-N | |
2,2-dimethoxyethylbenzene | |
60995 | |
98% |
98% | |
1mg KOH/g max. | |
1.0040g/mL | |
83°C | |
97.5% min. (GC) | |
C10H14O2 | |
C6H5CH2CH(OCH3)2 | |
100 g | |
1.004 | |
Solubility in water: insoluble. Other solubilities: soluble in 2 parts of 70% alcohol | |
COC(CC1=CC=CC=C1)OC | |
166.22 | |
166.22 | |
Liquid |
Chemical Identifiers
101-48-4 | |
MFCD00008487 | |
2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal | |
2,2-dimethoxyethylbenzene |
166.22 | |
WNJSKZBEWNVKGU-UHFFFAOYSA-N | |
60995 | |
COC(CC1=CC=CC=C1)OC |
Safety and Handling
EINECSNumber : 202-945-6
RTECSNumber : AB3040000
TSCA : TSCA
RUO – Research Use Only