Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenylacetaldehyde dimethyl acetal, 98%

$35.27 - $85.74

Chemical Identifiers

Chemical Name or Material Phenylacetaldehyde dimethyl acetal
CAS 101-48-4
Molecular Formula C10H14O2
Formula Weight 166.22
MDL Number MFCD00008487
Beilstein 879360
Synonym (2,2-Dimethoxyethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1873022

  SDS
alfa aesar™
A1873022
100g Each for $35.27

AAA1873036

  SDS
alfa aesar™
A1873036
500g Each for $85.74
Description & Specifications

Specifications

Formula Weight 166.22
Chemical Name or Material Phenylacetaldehyde dimethyl acetal
TSCA TSCA
Assay 98%
Molecular Formula C10H14O2
MDL Number MFCD00008487
CAS 101-48-4
Beilstein 879360
Synonym (2,2-Dimethoxyethyl)benzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Boiling Point 220°C
Flash Point 83°C (181°F)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AB3040000
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.