missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylacetaldehyde, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1426336

 View more versions of this product

Catalog No. AAA1426336



Description

Description

Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Phenylacetaldehyde
1.075
68°C (154°F)
1.535
500 g
Air sensitive
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
DTUQWGWMVIHBKE-UHFFFAOYSA-N
2-phenylacetaldehyde
998
120.15
122-78-1
194°C to 196°C
C8H8O
MFCD00006993
385791
14,7265
Miscible with alcohol and diethyl ether. Slightly miscible with water.
O=CCC1=CC=CC=C1
120.15
CHEBI:16424
95%

Chemical Identifiers

122-78-1
120.15
DTUQWGWMVIHBKE-UHFFFAOYSA-N
998
2-phenylacetaldehyde
C8H8O
MFCD00006993
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
CHEBI:16424
O=CCC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c

H227-H302-H317

EINECSNumber : 204-574-5

RTECSNumber : CY1420000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only