Learn More
Phenyl vinyl sulfone, 99+%, Thermo Scientific Chemicals
$55.74 - $605.07
Chemical Identifiers
CAS | 5535-48-8 |
---|---|
Molecular Formula | C8H8O2S |
Molecular Weight (g/mol) | 168.21 |
MDL Number | MFCD00007554 |
InChI Key | UJTPZISIAWDGFF-UHFFFAOYSA-N |
Synonym | phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon |
PubChem CID | 79664 |
IUPAC Name | ethenylsulfonylbenzene |
SMILES | C=CS(=O)(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1479403
|
Thermo Scientific Chemicals
A1479403 |
1 g |
Each for $55.74
|
|
|||||
AAA1479406
|
Thermo Scientific Chemicals
A1479406 |
5 g |
Each for $184.03
|
|
|||||
AAA1479414
|
Thermo Scientific Chemicals
A1479414 |
25 g |
Each for $605.07
|
|
|||||
Description
Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
It is soluble in most common organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
5535-48-8 | |
168.21 | |
UJTPZISIAWDGFF-UHFFFAOYSA-N | |
79664 | |
C=CS(=O)(=O)C1=CC=CC=C1 |
C8H8O2S | |
MFCD00007554 | |
phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon | |
ethenylsulfonylbenzene |
Specifications
5535-48-8 | |
C8H8O2S | |
1 g | |
phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon | |
UJTPZISIAWDGFF-UHFFFAOYSA-N | |
ethenylsulfonylbenzene | |
79664 | |
≥99% |
67°C to 71°C | |
MFCD00007554 | |
1906894 | |
It is soluble in most common organic solvents. | |
C=CS(=O)(=O)C1=CC=CC=C1 | |
168.21 | |
168.22 | |
Phenyl vinyl sulfone |
Safety and Handling
GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H318-H335
EINECSNumber : 226-890-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only