missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl vinyl sulfone, 99+%

Manufacturer:  Alfa Aesar™ A1479414

 View more versions of this product

Catalog No. AAA1479414



Description

Description

Applications
Phenyl vinyl sulfone is used as building blocks in the synthesis of a number of organic substances, participate in polymerization reactions, nucleophilic addition reaction, and in cycloaddition reactions. It is also used in the used in the synthesis of reactive dyes, synthesis of textile fibers, in particular cellulose fibers such as cotton and linen. Other applications in the field of medicine includes as powerful inhibitors of certain enzymatic processes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
It is soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

Phenyl vinyl sulfone
25g
C8H8O2S
1906894
UJTPZISIAWDGFF-UHFFFAOYSA-N
ethenylsulfonylbenzene
79664
≥99%
67°C to 71°C
5535-48-8
MFCD00007554
phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
C=CS(=O)(=O)C1=CC=CC=C1
168.21
168.22

Chemical Identifiers

5535-48-8
168.21
UJTPZISIAWDGFF-UHFFFAOYSA-N
79664
C=CS(=O)(=O)C1=CC=CC=C1
C8H8O2S
MFCD00007554
phenyl vinyl sulfone, vinylsulfonyl benzene, ethenesulfonyl benzene, benzene, ethenylsulfonyl, sulfone, phenyl vinyl, vinylsulfonylbenzene, phenyl vinyl sulphone, ethenylsulfonyl benzene, ethenesulfonyl-benzene, phenyl-vinylsulfon
ethenylsulfonylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P280g-P262-P305+P351+P338
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Recommended Storage : 2° to 8°C

SDS Documents