missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl trifluoromethyl sulfide, 98%

Manufacturer:  Alfa Aesar™ L1011403

 View more versions of this product

Catalog No. AAL1011403



Specifications

Specifications

Phenyl trifluoromethyl sulfide
Unpleasant
1g
C7H5F3S
UN1993
phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, trifluoromethyl thio benzene, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
C1=CC=C(C=C1)SC(F)(F)F
178.172
178.18
37°C (98°F)
140°C to 142°C
456-56-4
MFCD00040839
2043134
YQQKTCBMKQQOSM-UHFFFAOYSA-N
trifluoromethylsulfanylbenzene
68011
98%

Chemical Identifiers

456-56-4
178.172
YQQKTCBMKQQOSM-UHFFFAOYSA-N
68011
C1=CC=C(C=C1)SC(F)(F)F
C7H5F3S
MFCD00040839
phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, trifluoromethyl thio benzene, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
trifluoromethylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents