Please login to your online account to display your discounted pricing

Phenyl trifluoroacetate, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  309450050

 View more versions of this product

Catalog No. AC309450050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 190.12
Quantity 5mL
Color Clear colorless
EINECS Number 207-911-4
TSCA TSCA
Density 1.2700g/mL
Packaging Glass bottle
Refractive Index 1.4185 to 1.4225 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00039232
Synonym Trifluoroacetic acid phenyl ester
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Phenyl trifluoroacetate
Molecular Formula C8H5F3O2
Linear Formula CF3CO2C6H5
CAS 500-73-2

Specifications

Chemical Name or Material Phenyl trifluoroacetate
Name Note 98%
CAS 500-73-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H5F3O2
Linear Formula CF3CO2C6H5
Formula Weight 190.12
MDL Number MFCD00039232
Synonym Trifluoroacetic acid phenyl ester
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents