missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl trifluoroacetate, 98%, ACROS Organics™

Chemical Identifiers

CAS 500-73-2
Assay Percent Range 98%
Molecular Formula C8H5F3O2
Linear Formula CF3CO2C6H5
Formula Weight 190.12
MDL Number MFCD00039232
Synonym Trifluoroacetic acid phenyl ester
Chemical Name or Material Phenyl trifluoroacetate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC309450050 SDS Thermo Fisher Scientific
309450050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC309450250 SDS Thermo Fisher Scientific
AC309450250
Glass Bottle 25mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

500-73-2
C8H5F3O2
190.12
Trifluoroacetic acid phenyl ester
≥97.5%
98%
CF3CO2C6H5
MFCD00039232
Phenyl trifluoroacetate, 98%

Specifications

1.2700g/mL
5mL
Authentic
1.4185 to 1.4225 (20°C, 589nm)
98%
CF3CO2C6H5
MFCD00039232
Phenyl trifluoroacetate, 98%
Clear colorless
500-73-2
Glass bottle
Liquid
C8H5F3O2
190.12
Trifluoroacetic acid phenyl ester
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-911-4

TSCA : TSCA