Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl trifluoroacetate, 98%

Chemical Identifiers

Chemical Name or Material Phenyl trifluoroacetate
CAS 500-73-2
Molecular Formula C6H5OCOCF3
Formula Weight 190.12
MDL Number MFCD00039232
UN Number UN3272
Synonym Trifluoroacetic acid phenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2130406

  SDS
alfa aesar™
B2130406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2130414

  SDS
alfa aesar™
B2130414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.12
Chemical Name or Material Phenyl trifluoroacetate
TSCA TSCA
Assay 98%
Molecular Formula C6H5OCOCF3
MDL Number MFCD00039232
CAS 500-73-2
UN Number UN3272
Synonym Trifluoroacetic acid phenyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Flash Point 52°C (127°F)
Boiling Point 146° to 147°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.