Please login to your online account to display your discounted pricing

Phenyl Sulfoxide 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130970250

 View more versions of this product

Catalog No. AC130970250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 202.27
Quantity 25g
Color White to off-white
Beilstein 06, 300
Fieser 01,348; 17,365
EINECS Number 213-415-9
TSCA TSCA
RTECS Number DA9185000
Melting Point 66° to 71°C
Water 0.5% max. (K.F.)
Chemical Name or Material Phenyl sulfoxide
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H10OS
Linear Formula (C6H5)2SO
MDL Number MFCD00002085
CAS 945-51-7
Infrared Spectrum Authentic
Synonym Diphenyl sulfoxide
Packaging Glass bottle

Specifications

Chemical Name or Material Phenyl sulfoxide
Name Note 97%
CAS 945-51-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H10OS
Linear Formula (C6H5)2SO
Formula Weight 202.27
MDL Number MFCD00002085
Synonym Diphenyl sulfoxide