Please login to your online account to display your discounted pricing

Phenyl Sulfone 97%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: fisher scientific acros  130960050

 View more versions of this product

Catalog No. AC130960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 218.28
Physical Form Crystalline Powder
Quantity 5g
Color White
Packaging Glass Bottle
Melting Point 125° to 129°C
TSCA TSCA
RTECS Number SX2400000
EINECS Number 204-853-1
Beilstein 06, 300
Merck Index 15, 3372
Synonym Diphenyl sulfone
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Name Note 97%
Linear Formula (C6H5)2SO2
MDL Number MFCD00007548
CAS 127-63-9
Assay Percent Range 97%
Chemical Name or Material Phenyl sulfone
Molecular Formula C12H10O2S

Specifications

Chemical Name or Material Phenyl sulfone
Name Note 97%
CAS 127-63-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H10O2S
Linear Formula (C6H5)2SO2
Formula Weight 218.28
MDL Number MFCD00007548
Synonym Diphenyl sulfone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents