missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl Stearate, TCI America™

$126.81 - $340.70

Chemical Identifiers

CAS 637-55-8
Molecular Formula C24H40O2
Molecular Weight (g/mol) 360.582
MDL Number MFCD00026663
InChI Key NUMNZKICGJJSHN-UHFFFAOYSA-N
Synonym phenyl stearate, octadecanoic acid, phenyl ester, stearic acid phenyl ester, stearic acid, phenyl ester, phenylstearate, acmc-209nhu, octadecanoicacid,phenylester, stearic acid, phenyl ester 8ci
PubChem CID 69477
IUPAC Name phenyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O000725G View Documents TCI America
O000725G
25g Each for $340.70
 
O00075G View Documents TCI America
O00075G
5g Each for $126.81
 
Specifications

Chemical Identifiers

637-55-8
360.582
NUMNZKICGJJSHN-UHFFFAOYSA-N
69477
CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1
C24H40O2
MFCD00026663
phenyl stearate, octadecanoic acid, phenyl ester, stearic acid phenyl ester, stearic acid, phenyl ester, phenylstearate, acmc-209nhu, octadecanoicacid,phenylester, stearic acid, phenyl ester 8ci
phenyl octadecanoate

Specifications

267°C
53°C
637-55-8
MFCD00026663
NUMNZKICGJJSHN-UHFFFAOYSA-N
phenyl octadecanoate
69477
Crystal-Powder at 20°C
Phenyl Stearate
White
25g
C24H40O2
phenyl stearate, octadecanoic acid, phenyl ester, stearic acid phenyl ester, stearic acid, phenyl ester, phenylstearate, acmc-209nhu, octadecanoicacid,phenylester, stearic acid, phenyl ester 8ci
CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1
360.582
360.58
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes