Learn More
Phenyl propargyl ether, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 392730250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenyl propargyl ether | |
1.0300g/mL | |
76°C | |
97% min. (GC) | |
C9H8O | |
C6H5OCH2CCH | |
25mL | |
phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl | |
C#CCOC1=CC=CC=C1 | |
132.16 | |
132.16 |
13610-02-1 | |
89.0°C to 90.0°C (14.0mmHg) | |
Authentic | |
Glass bottle | |
1.5340 to 1.5360 (20°C, 589nm) | |
MFCD00048107 | |
1.03 | |
AIQRJSXKXVZCJO-UHFFFAOYSA-N | |
prop-2-ynoxybenzene | |
83613 | |
97+% |
Chemical Identifiers
13610-02-1 | |
132.16 | |
AIQRJSXKXVZCJO-UHFFFAOYSA-N | |
83613 | |
C#CCOC1=CC=CC=C1 |
C9H8O | |
MFCD00048107 | |
phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl | |
prop-2-ynoxybenzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 237-095-5
RUO â Research Use Only