missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl phosphorodiamidate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 300440010

 View more versions of this product

Catalog No. AC300440010


 

Specifications Description & Specifications

Specifications

Phenyl phosphorodiamidate
1g
7450-69-3
96.0% min.
Powder and Flakes
C6H9N2O2P
MFCD00014767
White to gray
97%
Authentic
Glass bottle
97%
172.12
06, IV, 751

Chemical Identifiers

7450-69-3
C6H9N2O2P
MFCD00014767
97%
172.12
Phenyl phosphorodiamidate, 97%

Safety and Handling

SDS & Safety Documents

MOISTURE SENSITIVE

EINECSNumber : 231-218-6