missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl phosphorodiamidate, 97%, ACROS Organics™

$45.83 - $53.50

Chemical Identifiers

CAS 7450-69-3
Assay Percent Range 97%
Molecular Formula C6H9N2O2P
Formula Weight 172.12
MDL Number MFCD00014767
Chemical Name or Material Phenyl phosphorodiamidate, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC300440010 SDS Thermo Fisher Scientific
300440010
Glass bottle 1g Each for $45.83
AC300440050 SDS Thermo Fisher Scientific
300440050
Glass bottle 5g Each for $53.50
Specifications Description & Specifications

Chemical Identifiers

7450-69-3
C6H9N2O2P
MFCD00014767
97%
172.12
Phenyl phosphorodiamidate, 97%

Specifications

Powder and Flakes
1g
06, IV, 751
Glass bottle
7450-69-3
97%
Phenyl phosphorodiamidate, 97%
172.12
White to gray
Authentic
96.0% min.
MFCD00014767
C6H9N2O2P

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 231-218-6