Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl nicotinate, 99%

$42.50 - $391.76

Chemical Identifiers

Chemical Name or Material Phenyl nicotinate
CAS 3468-53-9
Molecular Formula C12H9NO2
Formula Weight 199.21
MDL Number MFCD00023581
Beilstein 140968
Synonym Nicotinic acid phenyl ester; Phenyl pyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2221906

  SDS
alfa aesar™
B2221906
5g Each for $42.50

AAB2221922

  SDS
alfa aesar™
B2221922
100g Each for $391.76
Description & Specifications

Specifications

Formula Weight 199.21
Melting Point 70° to 74°C
Quantity 5g
Health Hazard 1 Danger
TSCA TSCA
Molecular Formula C12H9NO2
Beilstein 140968
Chemical Name or Material Phenyl nicotinate
Boiling Point 186° to 188°C (20mmHg)
Synonym Nicotinic acid phenyl ester; Phenyl pyridine-3-carboxylate
Assay 99%
Health Hazard 2 GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 3468-53-9
MDL Number MFCD00023581

Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.