missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl Methanesulfonate, TCI America™

Manufacturer:  TCI America P185225G

 View more versions of this product

Catalog No. P185225G



Specifications

Specifications

Phenyl Methanesulfonate
White-Yellow
25g
≥98.0% (GC)
MFCD00095143
WXVUCMFEGJUVTN-UHFFFAOYSA-N
phenyl methanesulfonate
316170
Crystal-Powder at 20°C
279°C
61°C
16156-59-5
C7H8O3S
Methanesulfonic Acid Phenyl Ester
CS(=O)(=O)OC1=CC=CC=C1
172.198
172.20

Chemical Identifiers

16156-59-5
172.198
WXVUCMFEGJUVTN-UHFFFAOYSA-N
316170
C7H8O3S
MFCD00095143
Methanesulfonic Acid Phenyl Ester
Safety and Handling

Safety and Handling

TSCA : No

Documents