Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl methacrylate, 95%, stab.

Chemical Identifiers

Chemical Name or Material Phenyl methacrylate
CAS 2177-70-0
Name Note stabilized
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00048117
UN Number UN3272
Beilstein 1907503
Synonym Methacrylic acid phenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1197806

  SDS
alfa aesar™
L1197806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1197814

  SDS
alfa aesar™
L1197814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.19
Synonym Methacrylic acid phenyl ester
Health Hazard 2 GHS H Statement
H334-H226-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Chemical Name or Material Phenyl methacrylate
Assay 95%
MDL Number MFCD00048117
Name Note stabilized
TSCA TSCA
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Beilstein 1907503
Boiling Point 115° to 116°C (10mmHg)
Health Hazard 1 Danger
UN Number UN3272
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Flash Point 60°C (140°F)
Quantity 5g
CAS 2177-70-0
Molecular Formula C10H10O2

Applications
Phenyl methacrylate is used as thermoset acrylic resin, fiber finishing agent and comonomers of resin lens.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H334-H226-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.