missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl methacrylate, 95%, stab.

Manufacturer:  Alfa Aesar™ L1197814

 View more versions of this product

Catalog No. AAL1197814



Description

Description

Applications
Phenyl methacrylate is used as thermoset acrylic resin, fiber finishing agent and comonomers of resin lens.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Specifications

Phenyl methacrylate
115°C to 116°C (10mmHg)
stabilized
C10H10O2
UN3272
phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, phenyl methacrylate, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
CC(=C)C(=O)OC1=CC=CC=C1
162.188
162.19
60°C (140°F)
25g
2177-70-0
MFCD00048117
1907503
QIWKUEJZZCOPFV-UHFFFAOYSA-N
phenyl 2-methylprop-2-enoate
75121
95%

Chemical Identifiers

2177-70-0
162.188
QIWKUEJZZCOPFV-UHFFFAOYSA-N
75121
CC(=C)C(=O)OC1=CC=CC=C1
C10H10O2
MFCD00048117
phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, phenyl methacrylate, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
phenyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H226-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Documents