missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl isothiocyanate, 98%, ACROS Organics™

Manufacturer:  Acros Organics 160975000

 View more versions of this product

Catalog No. AC160975000



Specifications

Specifications

Phenyl isothiocyanate
87°C
Glass bottle
1.1288
221.0°C
-21.0°C
103-72-0
C7H5NS
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
C1=CC=C(C=C1)N=C=S
135.184
CHEBI:85103
Liquid
1.1288g/mL
Authentic
1.6490 to 1.6510
0.1% max.
Yellow
500g
98%
C6H5NCS
QKFJKGMPGYROCL-UHFFFAOYSA-N
isothiocyanatobenzene
7673
135.19
98%

Chemical Identifiers

103-72-0
135.184
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
CHEBI:85103
C1=CC=C(C=C1)N=C=S
C7H5NS
QKFJKGMPGYROCL-UHFFFAOYSA-N
7673
isothiocyanatobenzene
Safety and Handling

Safety and Handling

Danger

SDS Documents