missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl isothiocyanate, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1159606

 View more versions of this product

Catalog No. AAA1159606



Description

Description

Applications
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

Solubility
Soluble in alcohol, and ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Store at 4°C.
Specifications

Specifications

Phenyl isothiocyanate
Pungent
-21°C
103-72-0
MFCD00004798
471392
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
C1=CC=C(C=C1)N=C=S
135.184
CHEBI:85103
97%
87°C (188°F)
220°C to 221°C
5g
C7H5NS
UN2927
14,7297
QKFJKGMPGYROCL-UHFFFAOYSA-N
isothiocyanatobenzene
7673
135.19

Chemical Identifiers

103-72-0
135.184
QKFJKGMPGYROCL-UHFFFAOYSA-N
7673
isothiocyanatobenzene
C7H5NS
MFCD00004798
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
CHEBI:85103
C1=CC=C(C=C1)N=C=S
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H314-H318-H227-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : NX9275000

TSCA : TSCA

SDS Documents