Please login to your online account to display your discounted pricing

Phenyl Isocyanate 99%, ACROS Organics™

$92.66 - $92.66

Chemical Identifiers

Chemical Name or Material Phenyl isocyanate
Name Note 99%
CAS 103-71-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5NO
Linear Formula C6H5NCO
Formula Weight 119.12
MDL Number MFCD00001994
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130690010

  SDS
thermo fisher scientific
130690010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC130690050

  SDS
thermo fisher scientific
130690050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC130692500

  SDS
thermo fisher scientific
130692500
Glass bottle 250g Each for $92.66
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 119.12
Quantity 1kg
Color Clear colorless to slightly yellow
Beilstein 12, 437
Merck Index 15, 7407
EINECS Number 203-137-6
TSCA TSCA
RTECS Number DA3675000
Density 1.095g/mL
Fieser 01,842; 02,322; 04,378; 07,284; 08,396; 10,309; 12,386
Refractive Index 1.5350 to 1.5370 (20°C, 589nm)
Chloride 0.3% max. (hydrolizable) (titr.)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Chemical Name or Material Phenyl isocyanate
Packaging Glass bottle
Additional Information organic solvent (GC): < 1.0%
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H5NO
Linear Formula C6H5NCO
MDL Number MFCD00001994
CAS 103-71-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.