Please login to your online account to display your discounted pricing

Phenyl Isocyanate 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  130690050

 View more versions of this product

Catalog No. AC130690050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 119.12
Quantity 5g
Color Clear colorless to slightly yellow
RTECS Number DA3675000
TSCA TSCA
Density 1.095g/mL
EINECS Number 203-137-6
Merck Index 15, 7407
Fieser 01,842; 02,322; 04,378; 07,284; 08,396; 10,309; 12,386
Beilstein 12, 437
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Refractive Index 1.5350 to 1.5370 (20°C, 589nm)
Additional Information organic solvent (GC): < 1.0%
Chloride 0.3% max. (hydrolizable) (titr.)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Linear Formula C6H5NCO
Name Note 99%
Chemical Name or Material Phenyl isocyanate
Molecular Formula C7H5NO
CAS 103-71-9
MDL Number MFCD00001994

Specifications

Chemical Name or Material Phenyl isocyanate
Name Note 99%
CAS 103-71-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5NO
Linear Formula C6H5NCO
Formula Weight 119.12
MDL Number MFCD00001994
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents