Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl formate, 95%

$67.05 - $235.05

Chemical Identifiers

Chemical Name or Material Phenyl formate
CAS 1864-94-4
Molecular Formula HCOOC6H5
Formula Weight 122.12
MDL Number MFCD00014126
Beilstein 507113
Synonym Formic acid phenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0028906

  SDS
alfa aesar™
L0028906
5g Each for $67.05

AAL0028914

  SDS
alfa aesar™
L0028914
25g Each for $235.05
Description & Specifications

Specifications

Formula Weight 122.12
Molecular Formula HCOOC6H5
CAS 1864-94-4
Chemical Name or Material Phenyl formate
Synonym Formic acid phenyl ester
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Assay 95%
Health Hazard 1 Warning
Beilstein 507113
Quantity 5g
Boiling Point 72° to 74°C (13mmHg)
Flash Point 71°C (159°F)
MDL Number MFCD00014126

Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

Solubility
Immiscible with water, alcohol, ether and benzene.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.