Learn More
Phenyl disulfide, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 173712500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenyl disulfide | |
882-33-7 | |
Authentic | |
Plastic bottle | |
(C6H5S-)2 | |
250g | |
05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156 | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
(phenyldisulfanyl)benzene | |
13436 | |
99% |
99% | |
191°C to 192°C (15.0mmHg) | |
98.5% min. (HPLC) | |
C12H10S2 | |
MFCD00003065 | |
06, 323 | |
15, 3355 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 | |
218.33 | |
218.33 |
Chemical Identifiers
882-33-7 | |
218.33 | |
GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
13436 | |
C1=CC=C(C=C1)SSC2=CC=CC=C2 |
C12H10S2 | |
MFCD00003065 | |
diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
(phenyldisulfanyl)benzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 212-926-4
RTECSNumber : SS6825000
TSCA : TSCA
RUO â Research Use Only