Learn More
Phenyl dichlorophosphate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130480250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenyl dichlorophosphate | |
770-12-7 | |
Brown to Colorless | |
241°C to 243°C | |
Authentic | |
Glass bottle | |
1.5210 to 1.523 | |
MFCD00002067 | |
06, 179 | |
1.412 | |
phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester | |
TXFOLHZMICYNRM-UHFFFAOYSA-N | |
dichlorophosphoryloxybenzene | |
13038 | |
99% |
99% | |
-1°C | |
1.4120g/mL | |
>110°C | |
98.5% min. (GC) | |
C6H5Cl2O2P | |
C6H5OP(O)Cl2 | |
25 g | |
01,847; 11,410; 12,384; 13,240; 15,146 | |
15, 7394 | |
Solubility in water: decomposes | |
C1=CC=C(C=C1)OP(=O)(Cl)Cl | |
210.98 | |
210.98 | |
Liquid |
Chemical Identifiers
770-12-7 | |
210.98 | |
TXFOLHZMICYNRM-UHFFFAOYSA-N | |
13038 | |
C1=CC=C(C=C1)OP(=O)(Cl)Cl |
C6H5Cl2O2P | |
MFCD00002067 | |
phenyl dichlorophosphate, phenyl phosphorodichloridate, dichlorophenoxyphosphine oxide, phosphorodichloridic acid, phenyl ester, phenylphosphoric dichloride, phenyl phosphorodichlorodate, phenoxydichlorophosphine oxide, phenyldichlorophosphate, phenoxyphosphoryl dichloride, phosphoroyl dichloride phenyl ester | |
dichlorophosphoryloxybenzene |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 212-220-6
RTECSNumber : TD4393000
TSCA : TSCA
RUO – Research Use Only