missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl carbamate, 98+%

Manufacturer:  Alfa Aesar™ L0094414

 View more versions of this product

Catalog No. AAL0094414



Description

Description

Applications
Phenyl carbamate, is used as various phenyl carbamate esters derived from amino acids, a dipeptide and amino acid esters and amides were prepared and assessed as potential prodrugs with the aim of protecting phenolic drugs against first-pass metabolism following per oral administration.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

149°C to 152°C
622-46-8
MFCD00007961
carbamic acid phenyl ester, carbamic acid, phenyl ester, ccris 5071, mono-phenol carbamate, mono-phenylcarbamate, o-phenyl carbamate, phenol carbamate, phenyl aminooate, phenyl carbamate, unii-jkb257u27v
BSCCSDNZEIHXOK-UHFFFAOYSA-N
phenyl carbamate
69322
≥98%
25g
C7H7NO2
2042627
Soluble in water.
C1=CC=C(C=C1)OC(=O)N
137.138
137.14

Chemical Identifiers

622-46-8
137.138
BSCCSDNZEIHXOK-UHFFFAOYSA-N
69322
C1=CC=C(C=C1)OC(=O)N
C7H7NO2
MFCD00007961
carbamic acid phenyl ester, carbamic acid, phenyl ester, ccris 5071, mono-phenol carbamate, mono-phenylcarbamate, o-phenyl carbamate, phenol carbamate, phenyl aminooate, phenyl carbamate, unii-jkb257u27v
phenyl carbamate
Safety and Handling

Safety and Handling

EINECSNumber : 210-737-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents