Please login to your online account to display your discounted pricing

γ-Phenyl-γ-butyrolactone, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  175210250

 View more versions of this product

Catalog No. AC175210250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 162.19
Physical Form Liquid After Melting
Quantity 25g
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 213-761-0
RTECS Number LU3690000
Density 1.1500g/mL
Synonym 4, 5-Dihydro-5-phenyl-2(3H)-furanone
Refractive Index 1.5415 to 1.5435 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
CAS 1008-76-0
Molecular Formula C10H10O2
Chemical Name or Material γ-Phenyl-γ-butyrolactone
MDL Number MFCD00005398
Assay Percent Range 97%

Specifications

Chemical Name or Material γ-Phenyl-γ-butyrolactone
Name Note 97%
CAS 1008-76-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00005398
Synonym 4, 5-Dihydro-5-phenyl-2(3H)-furanone